Assay Details
| Gene Symbol: Rnf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646690.1 | XP_007644880.1 | 6 - 7 | 983 | 59 | |
| XM_007646691.1 | XP_007644881.1 | 6 - 7 | 868 | 59 | ||
| GenBank mRNA | JI883398.1 | - | 5 - 6 | 793 | 59 | |
| JP045822.1 | - | 5 - 6 | 793 | 59 | ||
More Information
Set Membership: |
|

Rnf41
Genomic Map