Assay Details
| Gene Symbol: Sumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612006.1 | XP_007610196.1 | 5 - 6 | 330 | 105 | |
| GenBank mRNA | JP048009.1 | - | 1 - 2 | 370 | 105 | |
More Information
Set Membership: |
|

Sumo1
Genomic Map