Assay Details
| Gene Symbol: Rimkla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642478.1 | XP_007640668.1 | 6 - 7 | 773 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rimkla| Gene Symbol: Rimkla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642478.1 | XP_007640668.1 | 6 - 7 | 773 | 70 | |
Set Membership: |
|