Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654916.1 | XP_007653106.1 | 9 - 10 | 2185 | 63 | |
| GenBank mRNA | JI883768.1 | - | 10 - 11 | 2050 | 63 | |
| JP061251.1 | - | 10 - 11 | 2050 | 63 | ||
More Information
Set Membership: |
|

Spag5
Genomic Map