Assay Details
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647260.1 | XP_007645450.1 | 4 - 5 | 630 | 65 | |
XM_007647261.1 | XP_007645451.1 | 3 - 4 | 322 | 65 | ||
GenBank mRNA | JI873577.1 | - | 2 - 3 | 303 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
