Assay Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651545.1 | XP_007649735.1 | 4 - 5 | 555 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mybphl| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651545.1 | XP_007649735.1 | 4 - 5 | 555 | 58 | |
Set Membership: |
|