Assay Details
| Gene Symbol: Slc25a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642454.1 | XP_007640644.1 | 2 - 3 | 209 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a23| Gene Symbol: Slc25a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642454.1 | XP_007640644.1 | 2 - 3 | 209 | 68 | |
Set Membership: |
|