Assay Details
| Gene Symbol: LOC100755848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501142.2 | XP_003501190.1 | 40 - 41 | 5076 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100755848| Gene Symbol: LOC100755848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501142.2 | XP_003501190.1 | 40 - 41 | 5076 | 102 | |
Set Membership: |
|