Assay Details
| Gene Symbol: Ptges3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506214.2 | XP_003506262.2 | 7 - 8 | 482 | 68 | |
| XM_007646654.1 | XP_007644844.1 | 7 - 8 | 489 | 68 | ||
| GenBank mRNA | JI882749.1 | - | 6 - 7 | 532 | 68 | |
More Information
Set Membership: |
|

Ptges3
Genomic Map