Assay Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509676.2 | XP_003509724.1 | 13 - 14 | 1921 | 59 | |
| GenBank mRNA | JI864902.1 | - | 4 - 5 | 571 | 59 | |
| JP065708.1 | - | 4 - 5 | 570 | 59 | ||
More Information
Set Membership: |
|

Pdpr
Genomic Map