Assay Details
| Gene Symbol: LOC100774731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514033.2 | XP_003514081.2 | 5 - 6 | 1003 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100774731| Gene Symbol: LOC100774731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514033.2 | XP_003514081.2 | 5 - 6 | 1003 | 84 | |