Assay Details
| Gene Symbol: Vpreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643452.1 | XP_007641642.1 | 3 - 4 | 572 | 75 | |
| XM_007643453.1 | XP_007641643.1 | 3 - 4 | 575 | 75 | ||
| XM_007643454.1 | XP_007641644.1 | 2 - 3 | 459 | 75 | ||
More Information
Set Membership: |
|

Vpreb1
Genomic Map