Assay Details
| Gene Symbol: Vamp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505630.2 | XP_003505678.1 | 2 - 3 | 287 | 97 | |
| GenBank mRNA | JI888596.1 | - | 2 - 3 | 260 | 97 | |
| JP049909.1 | - | 2 - 3 | 260 | 97 | ||
More Information
Set Membership: |
|

Vamp8
Genomic Map