Assay Details
| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648399.1 | XP_007646589.1 | 5 - 6 | 589 | 65 | |
| GenBank mRNA | JI870756.1 | - | 471 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh10| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648399.1 | XP_007646589.1 | 5 - 6 | 589 | 65 | |
| GenBank mRNA | JI870756.1 | - | 471 | 65 | ||
Set Membership: |
|