Assay Details
| Gene Symbol: Iqcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641399.1 | XP_007639589.1 | 4 - 5 | 576 | 83 | |
| GenBank mRNA | JI892071.1 | - | 4 - 5 | 575 | 83 | |
More Information
Set Membership: |
|

Iqcc
Genomic Map