Assay Details
| Gene Symbol: LOC100773073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495320.2 | XP_003495368.1 | 4 - 5 | 413 | 75 | |
| GenBank mRNA | JI886619.1 | - | 4 - 5 | 432 | 75 | |
More Information
Set Membership: |
|

LOC100773073
Genomic Map