Assay Details
| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499338.2 | XP_003499386.1 | 1 - 2 | 108 | 66 | |
| GenBank mRNA | JI889120.1 | - | 1 - 2 | 66 | ||
More Information
Set Membership: |
|

Aph1b
Genomic Map