Assay Details
| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499338.2 | XP_003499386.1 | 2 - 3 | 285 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aph1b| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499338.2 | XP_003499386.1 | 2 - 3 | 285 | 72 | |
Set Membership: |
|