Assay Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499466.2 | XP_003499514.1 | 7 - 8 | 1963 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy1a3| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499466.2 | XP_003499514.1 | 7 - 8 | 1963 | 74 | |
Set Membership: |
|