Assay Details
| Gene Symbol: LOC100751546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI895270.1 | - | 3 - 4 | 290 | 94 | |
| JP058551.1 | - | 3 - 4 | 290 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751546| Gene Symbol: LOC100751546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI895270.1 | - | 3 - 4 | 290 | 94 | |
| JP058551.1 | - | 3 - 4 | 290 | 94 | ||