Assay Details
| Gene Symbol: LOC100751546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI895270.1 | - | 8 - 9 | 994 | 110 | |
| JP058551.1 | - | 8 - 9 | 994 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751546| Gene Symbol: LOC100751546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI895270.1 | - | 8 - 9 | 994 | 110 | |
| JP058551.1 | - | 8 - 9 | 994 | 110 | ||
Set Membership: |
|