Assay Details
| Gene Symbol: LOC100751546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI869690.1 | - | 4 - 5 | 613 | 91 | |
| JP048599.1 | - | 4 - 5 | 613 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751546| Gene Symbol: LOC100751546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI869690.1 | - | 4 - 5 | 613 | 91 | |
| JP048599.1 | - | 4 - 5 | 613 | 91 | ||