Assay Details
| Gene Symbol: Efna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501649.2 | XP_003501697.1 | 3 - 4 | 430 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efna4| Gene Symbol: Efna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501649.2 | XP_003501697.1 | 3 - 4 | 430 | 62 | |
Set Membership: |
|