Assay Details
| Gene Symbol: Mrpl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501672.2 | XP_003501720.1 | 2 - 3 | 115 | 74 | |
| XM_007642337.1 | XP_007640527.1 | 2 - 3 | 114 | 74 | ||
More Information
Set Membership: |
|

Mrpl14
Genomic Map