Assay Details
| Gene Symbol: Cdca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502244.2 | XP_003502292.1 | 2 - 3 | 475 | 59 | |
| GenBank mRNA | JI866018.1 | - | 2 - 3 | 458 | 59 | |
| JP048821.1 | - | 2 - 3 | 458 | 59 | ||
More Information
Set Membership: |
|

Cdca8
Genomic Map