Assay Details
| Gene Symbol: Glis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506306.2 | XP_003506354.1 | 6 - 7 | 1871 | 59 | |
| XM_007646813.1 | XP_007645003.1 | 7 - 8 | 2271 | 59 | ||
| XM_007646814.1 | XP_007645004.1 | 7 - 8 | 3211 | 59 | ||
| XM_007646815.1 | XP_007645005.1 | 6 - 7 | 1826 | 59 | ||
More Information
Set Membership: |
|

Glis3
Genomic Map