Assay Details
| Gene Symbol: Dctpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515071.2 | XP_003515119.1 | 1 - 2 | 172 | 70 | |
| GenBank mRNA | JI886586.1 | - | 1 - 2 | 79 | ||
More Information
Set Membership: |
|

Dctpp1
Genomic Map