Assay Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499695.2 | XP_003499743.1 | 3 - 4 | 337 | 106 | |
XM_007640424.1 | XP_007638614.1 | 3 - 4 | 320 | 106 | ||
XM_007640425.1 | XP_007638615.1 | 3 - 4 | 337 | 106 | ||
XM_007640426.1 | XP_007638616.1 | 3 - 4 | 337 | 106 | ||
GenBank mRNA | JP038886.1 | - | 3 - 4 | 331 | 106 |

More Information
Set Membership: |
![]() ![]() |
