Assay Details
| Gene Symbol: Tnfrsf12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508714.2 | XP_003508762.1 | 3 - 4 | 380 | 59 | |
| GenBank mRNA | JI889141.1 | - | 3 - 4 | 377 | 59 | |
| JP040392.1 | - | 3 - 4 | 377 | 59 | ||
More Information
Set Membership: |
|

Tnfrsf12a
Genomic Map