Assay Details
| Gene Symbol: LOC100763252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512291.2 | XP_003512339.1 | 3 - 4 | 343 | 85 | |
| XM_007652812.1 | XP_007651002.1 | 3 - 4 | 343 | 85 | ||
More Information
Set Membership: |
|

LOC100763252
Genomic Map