Assay Details
| Gene Symbol: Mxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648582.1 | XP_007646772.1 | 1 - 2 | 241 | 69 | |
| XM_007648583.1 | XP_007646773.1 | 1 - 2 | 241 | 69 | ||
More Information
Set Membership: |
|

Mxd1
Genomic Map