Assay Details
| Gene Symbol: Spsb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495496.2 | XP_003495544.1 | 3 - 4 | 1155 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spsb4| Gene Symbol: Spsb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495496.2 | XP_003495544.1 | 3 - 4 | 1155 | 81 | |
Set Membership: |
|