Assay Details
| Gene Symbol: Blvra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497812.2 | XP_003497860.1 | 7 - 8 | 690 | 57 | |
| GenBank mRNA | JI872457.1 | - | 7 - 8 | 690 | 57 | |
More Information
Set Membership: |
|

Blvra
Genomic Map