Assay Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502594.2 | XP_003502642.2 | 2 - 3 | 284 | 96 | |
| XM_007643171.1 | XP_007641361.1 | 2 - 3 | 273 | 96 | ||
| GenBank mRNA | JI875470.1 | - | 2 - 3 | 254 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007643171.1 transcripts.
Set Membership: |
|

Ppap2c
Genomic Map