Assay Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502594.2 | XP_003502642.2 | 4 - 5 | 624 | 75 | |
| XM_007643171.1 | XP_007641361.1 | 4 - 5 | 613 | 75 | ||
| GenBank mRNA | JI875470.1 | - | 4 - 5 | 594 | 75 | |
More Information
Set Membership: |
|

Ppap2c
Genomic Map