Assay Details
| Gene Symbol: LOC100763208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503700.1 | XP_003503748.1 | 1 - 2 | 92 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763208| Gene Symbol: LOC100763208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503700.1 | XP_003503748.1 | 1 - 2 | 92 | 77 | |