Assay Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505511.2 | XP_003505559.2 | 3 - 4 | 397 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp37| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505511.2 | XP_003505559.2 | 3 - 4 | 397 | 101 | |
Set Membership: |
|