Assay Details
| Gene Symbol: Mfsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507652.2 | XP_003507700.1 | 2 - 3 | 1034 | 80 | |
| XM_007648223.1 | XP_007646413.1 | 2 - 3 | 1034 | 80 | ||
| XM_007648224.1 | XP_007646414.1 | 2 - 3 | 1034 | 80 | ||
| XM_007648225.1 | XP_007646415.1 | 3 - 4 | 845 | 80 | ||
More Information
Set Membership: |
|

Mfsd3
Genomic Map