Assay Details
| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508066.2 | XP_003508114.1 | 6 - 7 | 943 | 60 | |
| GenBank mRNA | JI891110.1 | - | 3 - 4 | 473 | 60 | |
| JP055323.1 | - | 3 - 4 | 473 | 60 | ||
More Information
Set Membership: |
|

Pdlim1
Genomic Map