Assay Details
| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515973.2 | XP_003516021.2 | 1 - 2 | 436 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt28| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515973.2 | XP_003516021.2 | 1 - 2 | 436 | 65 | |
Set Membership: |
|