Assay Details
| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648004.1 | XP_007646194.1 | 3 - 4 | 375 | 72 | |
| XM_007648005.1 | XP_007646195.1 | 3 - 4 | 363 | 72 | ||
| GenBank mRNA | JP064485.1 | - | 3 - 4 | 350 | 72 | |
More Information
Set Membership: |
|

Znrd1
Genomic Map