Assay Details
| Gene Symbol: Znrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648004.1 | XP_007646194.1 | 4 - 5 | 484 | 96 | |
| XM_007648005.1 | XP_007646195.1 | 4 - 5 | 472 | 96 | ||
More Information
Set Membership: |
|

Znrd1
Genomic Map