Assay Details
| Gene Symbol: Trappc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649668.1 | XP_007647858.1 | 5 - 6 | 516 | 115 | |
| XM_007649669.1 | XP_007647859.1 | 5 - 6 | 641 | 115 | ||
| GenBank mRNA | JI881607.1 | - | 5 - 6 | 505 | 115 | |
More Information
Set Membership: |
|

Trappc2
Genomic Map