Assay Details
| Gene Symbol: LOC100763955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501368.1 | XP_003501416.1 | 2 - 3 | 907 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763955| Gene Symbol: LOC100763955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501368.1 | XP_003501416.1 | 2 - 3 | 907 | 76 | |
Set Membership: |
|