Assay Details
| Gene Symbol: Siah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502890.2 | XP_003502938.1 | 2 - 3 | 76 | 94 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siah2| Gene Symbol: Siah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502890.2 | XP_003502938.1 | 2 - 3 | 76 | 94 | |