Assay Details
| Gene Symbol: Mapk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505367.2 | XP_003505415.1 | 8 - 9 | 1045 | 68 | |
| XM_003505368.2 | XP_003505416.1 | 8 - 9 | 1045 | 68 | ||
| XM_007645898.1 | XP_007644088.1 | 9 - 10 | 1186 | 68 | ||
| XM_007645899.1 | XP_007644089.1 | 8 - 9 | 1045 | 68 | ||
| GenBank mRNA | JI880981.1 | - | 8 - 9 | 1065 | 68 | |
More Information
Set Membership: |
|

Mapk9
Genomic Map