Assay Details
| Gene Symbol: Asrgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508743.2 | XP_003508791.1 | 3 - 4 | 513 | 63 | |
| GenBank mRNA | JP047196.1 | - | 3 - 4 | 483 | 63 | |
More Information
Set Membership: |
|

Asrgl1
Genomic Map