Assay Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510310.2 | XP_003510358.1 | 4 - 5 | 547 | 76 | |
| GenBank mRNA | JI888696.1 | - | 2 - 3 | 96 | 76 | |
| JP051053.1 | - | 2 - 3 | 96 | 76 | ||
More Information
Set Membership: |
|

Pten
Genomic Map