Assay Details
| Gene Symbol: Atp5g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647788.1 | XP_007645978.1 | 4 - 5 | 451 | 94 | |
| GenBank mRNA | JI873659.1 | - | 3 - 4 | 188 | 94 | |
More Information
Set Membership: |
|

Atp5g2
Genomic Map