Assay Details
| Gene Symbol: LOC100759179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651568.1 | XP_007649758.1 | 8 - 9 | 1146 | 78 | |
| GenBank mRNA | JI888691.1 | - | 5 - 6 | 697 | 78 | |
| JP060538.1 | - | 5 - 6 | 696 | 78 | ||

LOC100759179
Genomic Map